missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19828-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.Specifications
HADH | |
Polyclonal | |
Unconjugated | |
HADH | |
HAD, HADH, HADH1, HADHSC, HCDH, HHF4, M/SCHAD, SCHAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113965, 15107, 3033 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16836, Q61425, Q9WVK7 | |
Hadh | |
HADH Fusion Protein Ag13928 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |