Learn More
Bochem™ HACCP Polyethylene Cutting Boards
Product Code. 12631915
Change view
Click to view available options
Dimensions (L x W x H):
610 x 460 x 25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12631915 610 x 460 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12631915 Supplier Bochem™ Supplier No. 12616
Estimated Shipment: 09-12-2026
Specifications
Color White
Material Polyethylene
Dimensions (L x W x H) 610 x 460 x 25 mm
Length (Metric) 610 mm
Width (Metric) 460 mm
Height (Metric) 25 mm