Learn More
Hanna Instruments™ HACCP-Compliant Salinity Foodcare Meter
Product Code. 11692550
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
11692550 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11692550 Supplier Hanna Instruments™ Supplier No. HI931102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Auto-ranges from sample to sample over a broad range without recalibration. Hanna Instruments™ HACCP-Compliant Salinity Foodcare Meter is a powerful meter that uses a sodium ion selective electrode (sold separately) to measure and clearly display the sodium content of a solution.
The HI931102 is an ion selective meter that has four ranges, capable of measuring concentrations from 0.150 to 300g/L. The HI931102 uses the FC300B combination sodium ISE (sold separately) to measure sodium. The calibration process is automatic at two points: the first at 3.00g/L and the second either at 0.30g/L (low range) or at 30.0g/L (high range).
  • Designed for use in food production
  • Provides grams NaCl/L or percent NaCl readings respectively through the MODE key
  • A separate temperature probe, HI7662, provides temperature readings from -20 to 120°C
  • A large and easy-to-read LCD shows sodium/sodium chloride and temperature values simultaneously
  • Onscreen instructions guide the user through the calibration procedure with easy-to-follow graphic symbols
Order Info
Note: this meter does not come with the FC300B Sodium Combination Ion Selective Electrode (ISE).
Specifications
Accuracy (ISE) ±5% of reading
Accuracy (Temperature) ±0.5
Battery Life 200 hr.
Battery Type 4 x 1.5V AA
Calibration Automatic at 3.00 g/L (HI7083) and either 0.30 g/L (HI7085) or 30.0 g/L (HI7081)
Certifications/Compliance HACCP
Description Salinity meter for use in food production
Calibration Points (ISE) 1 or 2
Range (ISE) 0.150 to 1.500 g/L, 1.50 to 15.00 g/L, 15.0 to 150.0 g/L, 150 to 300 g/L, 0.00 to 30.00% NaCl
Resolution (ISE) 0.001 g/L, 0.01 g/L, 0.1 g/L, 1 g/L, 0.01% NaCl
Temperature (Metric) Range 0.0°C to 80.0°C, compensation fixed at 25°C
Display Type LCD
Includes 4 x 1.5V AA batteries and rugged carrying case (probes not included)
Probe Type Required: FC300B glass combination sodium-sensitive electrode, Optional: HI7662 temperature probe (both sold separately)
Relative Humidity Range 100%
Resolution (Temperature) 0.1
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Dimensions (L x W x H) 196 x 80 x 60 mm (7.7 x 3.1 x 2.4 in.)
Weight (English) 15.9 oz.
Weight (Metric) 450 g