missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H3F3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10265-1-AP-20UL
This item is not returnable.
View return policy
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21. 3.Specifications
| H3F3B | |
| Polyclonal | |
| Unconjugated | |
| H3F3B | |
| H3 histone, family 3B (H3.3B), H3.3A, H3.3B, H3F3, H3F3A, H3F3B, Histone H3.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117056, 15081, 3021 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P84243, P84245 | |
| H3f3b | |
| H3F3B Fusion Protein Ag0326 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto