Learn More
H2AFV Antibody (2F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00094239-M08
This item is not returnable.
View return policy
Description
H2AFV Monoclonal antibody specifically detects H2AFV in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| H2AFV | |
| Monoclonal | |
| Unconjugated | |
| AAH00098 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 2F9 | |
| In 1x PBS, pH 7.4 | |
| FLJ26479, H2A histone family, member V, H2A.F/Z, H2AV, histone H2A.F/Z, histone H2A.V, MGC10170, MGC10831, MGC1947, purine-rich binding element protein B | |
| H2AFV (AAH00098, 1 a.a. ∽ 128 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAGGKAGRDSGKAKAKAVSRSQRAGLQFPVGRIHRHLKTRTTSHGRVGATAAVYSAAILEYLTAEVLELAGNASKDLKVKRITPRHLQLAIRGDEELDSLIKATIAGGGVIPHIHKSLIGKKGQQKTA | |
| 0.1 mg | |
| Direct Reversal of DNA Damage | |
| 94239 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |