Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Product Code. p-7144721 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
18 x 15 mL Adapter
33 x 10/12 mL Adapter
400 mL Bottle/Centrilab Adapter
42 x 5/7 mL Adapter
8 x 50 mL Adapter
Biocontainment Lids
Buckets
Cross
Microplate Adapter
Replacement O-Rings
Unit Size:
Each
Pack of 2
Product Code. Description unitSize
16525311 8 x 50 mL Adapter Pack of 2
16535311 18 x 15 mL Adapter Pack of 2
16545311 33 x 10/12 mL Adapter Pack of 2
16555311 42 x 5/7 mL Adapter Pack of 2
16575311 400 mL Bottle/Centrilab Adapter Pack of 2
16595301 Cross Each
16505311 Buckets Pack of 2
16515311 Biocontainment Lids Pack of 2
16565311 Microplate Adapter Pack of 2
16585311 Replacement O-Rings Pack of 2
10 options
Product Code. 16525311
Brand: Thermo Scientific™ 75003303
Spin more with less! High capacity and flexibility—all in a small footprint.
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
  • Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Get results faster without the need to change hardware
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
TRUSTED_SUSTAINABILITY
Specifications
Description 8 x 50 mL Adapter
Material Molded Polyamide Nylone
Type Swinging Bucket Rotor
Capacity 8 x 50 mL Conical Each Adapter
Max. Speed 4297
For Use With (Equipment) Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Biocontainment Yes
Autoclavable Autoclavable
Quantity Set of 2