Learn More
Bel-Art™ H-B Durac Specific Gravity and Baume Dual Scale Hydrometer For Liquids Heavier Than Water
Description
- Used in the chemistry, pharmacology, food and beverage, and starch industries to name a few, these dual scale, plain form hydrometers measure specific gravity/relative density (g/cm3) of liquids lighter and heavier than water in various ranges from 0.700 to 2.00, and Baume in various ranges from 10° to 70°, or choose the Universal model to span all ranges.
- Lead free ballast and glass
- Individually serialized
- Temperature standardization: of 60°F (15°C)
- Hand inspected by our U.S. quality team
- Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
- Note: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Spécification
Spécification
| Type | Hydrometer |
| Includes | Statement of Accuracy |
| Material | Ballast, Glass |
| Ballast Type | Lead Free |
| Baume Range | 10° to 70° |
| For Use With (Application) | Measure Specific Gravity/Relative Density (g/cm3) of Liquids Lighter and Heavier Than Water |
| Specific Gravity Range | 0.700 to 2.00 g/cm3 |
| Temperature of Standardization | 15°C |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?