missing translation for 'onlineSavingsMsg'
Learn More

GYKI 53655 hydrochloride, MedChemExpress

Product Code. p-200099250
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493808

Brand: Medchem Express HY103228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GYKI 53655 (LY300168) hydrochloride is an α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) antagonist.

GYKI 53655 (LY300168) hydrochloride is an α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) antagonist.

Specifications

Chemical Name or Material GYKI 53655 hydrochloride
CAS 143692-48-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H21ClN4O3
Quantity 10 mg
Synonym LY300168 hydrochloride
Solubility Information DMSO : ≥ 160 mg/mL (411.47 mM) ∣H2O : 8 mg/mL (20.57 mM; Need ultrasonic and warming)
SMILES O=C(N1N=C(C2=CC=C(N)C=C2)C3=CC(OCO4)=C4C=C3CC1C)NC.[H]Cl
Molecular Weight (g/mol) 388.85
Formula Weight 388.85
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.