Learn More
GW788388, MedChemExpress
Product Code. 16444866
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16444866
Brand: Medchem Express HY10326
MedChemExpress GW788388 is a potent and selective inhibitor of ALK5 with IC50 of 18 nM, and also inhibits TGF-β type II receptor and activin type II receptor activities, without inhibiting BMP type II receptor.
GW788388 is a potent and selective inhibitor of ALK5 with IC50 of 18 nM, and also inhibits TGF-β type II receptor and activin type II receptor activities, without inhibiting BMP type II receptor.
Specifications
Chemical Name or Material GW788388
CAS 452342-67-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H23N5O2
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (58.76 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1)C2=NC=CC(C3=CNN=C3C4=NC=CC=C4)=C2)NC5CCOCC5
Molecular Weight (g/mol) 425.48
Formula Weight 425.48
Percent Purity 98.3%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.