Learn More
Notrax Guzzler™ Absorbent Barrier Carpet Mat
Product Code. 17721998
Click to view available options
Dimensions (L x W):
90 x 60 cm
120 x 90 cm
150 x 90 cm
300 x 90 cm
180 x 120 cm
300 x 120 cm
Unit Size:
Each
Product Code. 17721998
Brand: Notrax 166S0046CH
Only null left
Add to basket
Add to basket
Notrax Guzzler™ Tough, long-lasting carpet mat with raised rubber borders on all 4 sides acts as a containment barrier and can hold up to 6 liters of water per square meter. The raised waffle pattern works double duty to remove and trap dirt and moisture.
  • Tough, long-lasting carpet mat with raised rubber borders on all 4 sides acts as a containment barrier and can hold up to 6 liters of water per square meter
  • The raised waffle pattern works double duty to remove and trap dirt and moisture
  • Rubber reinforced borders and backing provide maximum strength
  • Cleated backing minimizes movement on all surfaces and keeps mat elevated allowing wet hard floors to dry underneath
Specifications
Dimensions (L x W) 180 x 120 cm
Color Charcoal
Length (English) 71 in.
Length (Metric) 180 cm
Material Polypropylene
Type Industrial and Commercial Matting
Width (Metric) 120 cm