missing translation for 'onlineSavingsMsg'
Learn More

Guggulsterone, MedChemExpress

Product Code. p-200098137
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480497

Brand: Medchem Express HY107738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guggulsterone is a plant sterol derived from the gum resin of the tree Commiphora wightii.

Guggulsterone inhibits the growth of a wide variety of tumor cells and induces apoptosis through down regulation of antiapoptotic gene products (IAP1, xIAP, Bfl-1/A1, Bcl-2, cFLIP and survivin), modulation of cell cycle proteins (cyclin D1 and c-Myc), activation of caspases and JNK, inhibition of Akt. Guggulsterone, a farnesoid X receptor (FXR) antagonist, decreases CDCA-induced FXR activation with IC50s of 17 and 15 μM for Z- and E-Guggulsterone, respectively.

Specifications

Chemical Name or Material Guggulsterone
Physical Form Solid
CAS 95975-55-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H28O2
For Use With (Application) Cancer-Kinase/protease
Synonym Z/E-Guggulsterone
Solubility Information DMSO : 20.83 mg/mL (66.67 mM; Need ultrasonic)
SMILES C[C@@]1(/C2=C/C)[C@](CC2=O)([H])[C@@]3([H])[C@]([C@@]4(C(CC3)=CC(CC4)=O)C)([H])CC1
Molecular Weight (g/mol) 312.45
Formula Weight 312.45
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.