Learn More
OHAUS™ Guardian 7000 Hotplate-Stirrer
Product Code. 16600492 Shop All Ohaus Products
Click to view available options
Model:
e-G71HS07C
e-G71HS10C
e-G71HSRDM
Unit Size:
Each
Product Code. 16600492
Brand: OHAUS™ 30500623
Only null left
Add to basket
Add to basket
Durable hotplate stirrers with superior safety features and intelligent performance
  • Protect your lab with patented SmartPresence™ & SmartLink™ technologies. These exclusive features automatically shut off the heater if no one is detected after a user set "time out" period has passed.
  • An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
  • Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
TRUSTED_SUSTAINABILITY
Specifications
Top Plate Material Aluminum
Voltage 230 V
Shipping Weight (Metric) 2.812 kg
Electrical Requirements 230 V, 4.6 A, 50/60 Hz
Model e-G71HSRDM
Length (English) 10.50 in.
Height (Metric) 126.000 mm
Ambient Temperature + 5°C – 380°C
Height (English) 5.00 in.
Frequency 50/60 Hz
Width (Metric) 173.00 mm
Length (Metric) 267.00 mm
Width (English) 6.80 in.
Weight (English) 5.700 lb
Weight (Metric) 2.600 kg
Plug Type EU