Learn More
OHAUS™ Guardian 5000 Hotplate-Stirrer
Product Code. 16680452 Shop All Ohaus Products
Change view
Click to view available options
Model:
e-G51HS07C
e-G51HS10C
e-G51HSRDM
Unit Size:
Each
Product Code. Model unitSize
16680452 e-G51HSRDM Each
16640452 e-G51HS07C Each
16670452 e-G51HS10C Each
3 options
Product Code. 16680452
Brand: OHAUS™ 30500563
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reliable hotplate stirrers with exceptional safety features for everyday use.
  • An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
  • Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
  • The bright LCD display includes large temperature and speed settings, intuitive icons and a prominent hot top indicator - all designed to quickly and easliy monitor performance from across the lab.
TRUSTED_SUSTAINABILITY
Specifications
Top Plate Material Aluminum
Voltage 230 V
Shipping Weight (Metric) 2.812 kg
Electrical Requirements 230 V, 4.6 A, 50/60 Hz
Model e-G51HSRDM
Length (English) 10.50 in.
Height (Metric) 126.000 mm
Ambient Temperature + 5°C – 380°C
Height (English) 5.00 in.
Frequency 50/60 Hz
Width (Metric) 173.00 mm
Length (Metric) 267.00 mm
Width (English) 6.80 in.
Weight (English) 5.700 lb
Weight (Metric) 2.600 kg
Plug Type EU