Learn More
Guanidinium Thiocyanate, Millipore™
Specifications
Specifications
| CAS | 593-84-0 |
| Melting Point | 118°C |
| Density | 1.29 g/cm3 at 20°C |
| Molecular Formula | H2NC(NH)NH2 · HSCN |
| MDL Number | MFCD00013027 |
| Quantity | 250 g |
| Synonym | Guanidinium thiocyanate, Guanidinium thiocyanate |
| Molecular Weight (g/mol) | 118.16 |
| Grade | Research |