Learn More
Scharlab GUANIDINHYDROKLORID 250G
Brand:  Scharlab GU00600250
GUANIDINHYDROKLORID 250G
Product Code. 11304164
  • ONLINE EXCLUSIVE PRICE
    1428.00 NOK
    874.89 NOK / 250g
    Save 553.11 NOK
    38% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Guanidine hydrochloride, synthesis grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .