missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, 99+%, MP Biomedicals
BIOPHARMA

Product Code. 11486551 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
3 kg
Unit Size:
100g
1kg
25g
3kg
500g
This item is not returnable. View return policy

Product Code. 11486551

Brand: MP Biomedicals 0219482690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Assay Percent Range 99+%
Quantity 500 g
Formula Weight 95.53
Chemical Name or Material Guanidine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.