Learn More
Guanidine Hydrobromide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 19244-98-5 |
| Molecular Formula | CH6BrN3 |
| MDL Number | MFCD03094062 |
| Quantity | 25 g |
| SMILES | NC(N)=N.[H]Br |
| Molecular Weight (g/mol) | 139.98 |
| Percent Purity | 98% |
| Chemical Name or Material | Guanidine Hydrobromide |