Learn More
Guaiacol-Beta-D-glucopyranoside-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Guaiacol-beta-D-glucopyranoside-d3 |
| CAS | 1208109-55-0 |
| Molecular Formula | C13H15D3O7 |
| Quantity | 1 mg |
| Synonym | 2-(Methoxy-d3)phenyl β-D-Glucopyranoside |
| InChI Formula | InChI=1S/C13H18O7/c1-18-7-4-2-3-5-8(7)19-13-12(17)11(16)10(15)9(6-14)20-13/h2-5,9-17H,6H2,1H3/t9-,10-,11+,12-,13-/m1/s1/i1D3 |
| SMILES | O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=C(OC([2H])([2H])[2H])C=CC=C2)[C@@H]1O |
| Molecular Weight (g/mol) | 289.3 |
| Formula Weight | 289.1241 |