missing translation for 'onlineSavingsMsg'
Learn More

Guadecitabine sodium, MedChemExpress

Product Code. p-200096052
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16456378

Brand: Medchem Express HY15229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guadecitabine sodium (SGI-110 sodium) is a second-generation DNA methyltransferases (DNMT) inhibitor for research of acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS).

Guadecitabine sodium (SGI-110 sodium) is a second-generation DNA methyltransferases (DNMT) inhibitor for research of acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS).

Specifications

Chemical Name or Material Guadecitabine sodium
CAS 929904-85-8
Color White
Purity Grade Notes Research
Molecular Formula C18H23N9NaO10P
Quantity 10 mg
Synonym SGI-110 sodium S-110 sodium
Solubility Information H2O : 50 mg/mL (86.30 mM; Need ultrasonic and warming) ∣DMSO : 50 mg/mL (86.30 mM; Need ultrasonic)
SMILES O=C1C2=C(N([C@H]3C[C@H](O)[C@@H](COP(O[C@@H]4[C@@H](CO)O[C@@H](N5C=NC(N)=NC5=O)C4)(O[Na])=O)O3)C=N2)NC(N)=N1
Molecular Weight (g/mol) 579.39
Formula Weight 579.39
Percent Purity 95.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.