Learn More
Guadecitabine sodium, MedChemExpress
Product Code. 16456378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16456378 10 mg 10mg
16448967 5 mg 5mg
2 options
Product Code. 16456378
Brand: Medchem Express HY15229
MedChemExpress Guadecitabine sodium (SGI-110 sodium) is a second-generation DNA methyltransferases (DNMT) inhibitor for research of acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS).
Guadecitabine sodium (SGI-110 sodium) is a second-generation DNA methyltransferases (DNMT) inhibitor for research of acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS).
Specifications
Chemical Name or Material Guadecitabine sodium
CAS 929904-85-8
Color White
Purity Grade Notes Research
Molecular Formula C18H23N9NaO10P
Quantity 10 mg
Synonym SGI-110 sodium S-110 sodium
Solubility Information H2O : 50 mg/mL (86.30 mM; Need ultrasonic and warming) ∣DMSO : 50 mg/mL (86.30 mM; Need ultrasonic)
SMILES O=C1C2=C(N([C@H]3C[C@H](O)[C@@H](COP(O[C@@H]4[C@@H](CO)O[C@@H](N5C=NC(N)=NC5=O)C4)(O[Na])=O)O3)C=N2)NC(N)=N1
Molecular Weight (g/mol) 579.39
Formula Weight 579.39
Percent Purity 95.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death