missing translation for 'onlineSavingsMsg'
Learn More

GTF2H1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16806383 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16806383 20 μL 20µL
16896373 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16806383 Supplier Proteintech Supplier No. 101731AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TFIIH, a multisubunit complex is involved in several biological fundamental mechanisms of the cell: transcription, nucleotide excision repair and cell cycle regulation. p62 is one of the six subunits that constitutes the core of TFIIH. Analysis of the expression of the p62 gene reveals an over expression in testis tissue. This subunit of TFIIH participates in a variety of protein interactions. For example, Rb competes with TBP and p62 for binding to E2F thus repressing E2F mediated transactivation; herpes simplex virus VP16 and human p53, directly interact with the p62 subunit of TFIIH. In addition, TFIIH, via p62 phosphorylation is the major target for mitotic inactivation of transcription. GTF2H1 is a component of the core GTF2H basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. GTF2H1 is one of the six subunits forming the core GTF2H basal transcription factor.

Specifications

Antigen GTF2H1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GTF2H1
Gene Accession No. P32780, Q9DBA9
Gene Alias BTF2, BTF2 p62, GTF2H1, TFB1, TFIIH
Gene Symbols Gtf2h1
Host Species Rabbit
Immunogen GTF2H1 Fusion Protein Ag0229
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14884, 2965
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.