missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21381-1-AP-150UL
This item is not returnable.
View return policy
Description
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.Specifications
| GTF2A1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2A1 | |
| GTF2A1, TF2A1, TFIIA, TFIIA 42, TFIIA p19 subunit, TFIIAL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2957, 83602, 83830 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08949, P52655, Q99PM3 | |
| GTF2A1 | |
| GTF2A1 Fusion Protein Ag15994 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction