missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14562-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).Specifications
GSTO2 | |
Polyclonal | |
Unconjugated | |
GSTO2 | |
GSTO 2, GSTO2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
119391, 309465, 68214 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AXV9, Q8K2Q2, Q9H4Y5 | |
Gsto2 | |
GSTO2 Fusion Protein Ag6120 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |