Learn More
Abnova™ GSTA4 (Human) Recombinant Protein
Product Code. p-7164310
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16125831
Brand: Abnova™ H00002941P01.10ug
Human GSTA4 full-length ORF ( AAH15523, 1 a.a. - 222 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAARPKLHYPNGRGRMESVRWVLAAAGVEFDEEFLETKEQLYKLQDGNHLLFQQVPMVEIDGMKLVQTRSILHYIADKHNLFGKNLKERTLIDMYVEGTLDLLELLIMHPFLKPDDQQKEVVNMAQKAIIRYFPVFEKILRGHGQSFLVGNQLSLADVILLQTILALEEKIPNILSAFPFLQEYTVKLSNIPTIKRFLEPGSKKKPPPDEIYVRTVYNIFRP
Specifications
Accession Number AAH15523
Gene ID (Entrez) 2941
Name glutathione S-transferase alpha 4
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp686D21185, GSTA4-4, GTA4
Gene Symbol GSTA4
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer