Learn More
Abnova™ GSTA1 (Human) Recombinant Protein
Product Code. p-7164308
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16115821
Brand: Abnova™ H00002938P01.10ug
Human GSTA1 full-length ORF ( AAH53578.1, 1 a.a. - 222 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAEKPKLHYFNARGRMESTRWLLAAAGVEFEEKFIKSAEDLDKLRNDGYLMFQQVPMVEIDGMKLVQTRAILNYIASKYNLYGKDIKERALIDMYIEGIADLGEMILLLPVCPPEEKDAKLALIKEKIKNRYFPAFEKVLKSHGQDYLVGNKLSRADIHLVELLYYVEELDSSLISSFPLLKALKTRISNLPTVKKFLQPGSPRKPPMDEKSLEEARKIFRF
Specifications
Accession Number AAH53578.1
Gene ID (Entrez) 2938
Name glutathione S-transferase alpha 1
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias GST2, GSTA1-1, GTH1, MGC131939
Gene Symbol GSTA1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer