Learn More
Kgw Isotherm™ GSS Stainless Steel Dewar Flasks
Product Code. 10672092
Change view
Click to view available options
Capacity (Metric):
500 mL
1 L
6 L
Height (Metric):
156 mm
180 mm
206 mm
270 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) unitSize
10672092 1 L 156 mm Each
10259090 1 L 206 mm Each
10548041 500 mL 180 mm Each
10510162 6 L 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10672092 Supplier Kgw Isotherm™ Supplier No. 2301
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 01-09-2026
Specifications
For Use With (Application) Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants
Height (Metric) 156 mm
Includes Loose Lid Insulating Plug
Capacity (Metric) 1 L
Diameter (Metric) Inner 100 mm
Material GSS Stainless Steel