missing translation for 'onlineSavingsMsg'
Learn More

GSK369796 Dihydrochloride, MedChemExpress

Product Code. 16451227
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451227

Brand: Medchem Express HY12082A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.

GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.

Specifications

Chemical Name or Material GSK369796 Dihydrochloride
Physical Form Solid
CAS 1010411-21-8
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H24Cl3N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (233.22 mM; Need ultrasonic) ∣H2O : 50 mg/mL (116.61 mM; Need ultrasonic)
SMILES OC1=CC(NC2=CC=NC3=CC(Cl)=CC=C23)=CC=C1CNC(C)(C)C.[H]Cl.[H]Cl
Molecular Weight (g/mol) 428.78
Formula Weight 428.78
Percent Purity 98.32%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.