Learn More
GSK360A, TRC
Product Code. 30133330
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133330 25 mg 25mg
30135005 10 mg 10mg
30004445 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133330 Supplier Toronto Research Chemicals Supplier No. TRCG79760025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 931399-19-8
Molecular Formula C17H17FN2O5
Molecular Weight (g/mol) 348.33
Synonym Glycine, N-[[1-(2-cyclopropylethyl)-6-fluoro-1,2-dihydro-4-hydroxy-2-oxo-3-quinolinyl]carbonyl]- (ACI), N-[[1-(2-Cyclopropylethyl)-6-fluoro-1,2-dihydro-4-hydroxy-2-oxo-3-quinolinyl]carbonyl]glycine (ACI), GSK 1120360A, N-[[1-(2-Cyclopropylethyl)-6-fluoro-4-hydroxy-2-oxo-1,2-dihydro-3-quinolinyl]carbonyl]glycine
IUPAC Name 2-[[1-(2-cyclopropylethyl)-6-fluoro-4-hydroxy-2-oxoquinoline-3-carbonyl]amino]acetic acid
SMILES OC(=O)CNC(=O)C1=C(O)c2cc(F)ccc2N(CCC3CC3)C1=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C17H17FN2O5/c18-10-3-4-12-11(7-10)15(23)14(16(24)19-8-13(21)22)17(25)20(12)6-5-9-1-2-9/h3-4,7,9,23H,1-2,5-6,8H2,(H,19,24)(H,21,22)
Formula Weight 348.1121
Percent Purity >95%
Chemical Name or Material GSK360A
Grade HPLC