Learn More
GSK2807 Trifluoroacetate, MedChemExpress
Product Code. 16410287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16410287 50 mg 50mg
16406647 5 mg 5mg
16404956 100 mg 100mg
16403928 10 mg 10mg
16402916 25 mg 25mg
5 options
Product Code. 16410287
Brand: Medchem Express HY104009A
MedChemExpress GSK2807 Trifluoroacetate is a potent, selective and SAM-competitive inhibitor of SMYD3, with a Ki of 14 nM and an IC50 of 130 nM.
GSK2807 Trifluoroacetate is a potent, selective and SAM-competitive inhibitor of SMYD3, with a Ki of 14 nM and an IC50 of 130 nM.
Specifications
Chemical Name or Material GSK2807 Trifluoroacetate
CAS 2245255-66-5
Color White
Purity Grade Notes Research
Molecular Formula C21H33F3N8O7
Quantity 50 mg
Solubility Information DMSO : 250 mg/mL (441.28 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (88.26 mM)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](CN(CC[C@H](N)C(O)=O)CCCN(C)C)O3)C=N2)=NC=N1.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 566.53
Formula Weight 566.53
Percent Purity 95.0%
Grade Research
Physical Form Solid