missing translation for 'onlineSavingsMsg'
Learn More

GSK137647A, MedChemExpress

Product Code. p-200100796
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16468008

Brand: Medchem Express HY19995

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK137647A (GSK 137647) is a potent, selective free fatty acid receptor 4 (FFA4) agonist with pEC50 values of 6.3, 6.2, and 6.1 for human, mouse and rat FFA4, and pEC50 values < 4.5 for all three species for FFA1, FFA2, and FFA3, respectively.

GSK137647A has anti-inflammatory activity. GSK137647A induces insulin secretion and inhibits epithelial ion transport, GSK137647A is related to regulation of glucose homeostasis and anti-inflammatory response.

Specifications

Chemical Name or Material GSK137647A
CAS 349085-82-1
Color White
Purity Grade Notes Research
Molecular Formula C16H19NO3S
Quantity 10 mg
Synonym GSK 137647
Solubility Information DMSO : ≥ 100 mg/mL (327.45 mM) ∣H2O : mg/mL (ultrasonic;warming) (Need ultrasonic and warming)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=C(C)C=C(C)C=C2C)=O
Molecular Weight (g/mol) 305.39
Formula Weight 305.39
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.