Learn More
GSK1059615, MedChemExpress
Product Code. 16487677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16487677 5 mg 5mg
16464938 10 mg 10mg
16445716 100 mg 100mg
16431187 50 mg 50mg
4 options
Product Code. 16487677
Brand: Medchem Express HY12036
MedChemExpress GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively.
GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively.
Specifications
Chemical Name or Material GSK1059615
Physical Form Solid
CAS 958852-01-2
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H11N3O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 5 mg/mL (15.00 mM; Need ultrasonic)
SMILES O=C(S/C1=C\C2=CC=C3N=CC=C(C3=C2)C4=CC=NC=C4)NC1=O
Molecular Weight (g/mol) 333.36
Formula Weight 333.36
Percent Purity 98.0%
Grade Research