missing translation for 'onlineSavingsMsg'
Learn More

GSDMD Rabbit anti-Human, Clone: 2N24, Proteintech

Product Code. 17669806 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17669806 20 μL 20µL
17659806 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17669806

Brand: Proteintech 809181RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

GSDMD antibody apecifically detetects GSDMD antigen in Human Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GSDMD
Applications Western Blot, ELISA
Clone 2N24
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC008904
Gene Symbols GSDMD
Host Species Rabbit
Immunogen GSDMD fusion protein Ag14757
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 79792
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.