missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMD Rabbit anti-Human, Clone: 2N24, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80918-1-RR-100UL
This item is not returnable.
View return policy
Description
GSDMD antibody apecifically detetects GSDMD antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
| GSDMD | |
| 2N24 | |
| Unconjugated | |
| BC008904 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| GSDMD | |
| GSDMD fusion protein Ag14757 | |
| 100 μL | |
| 79792 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction