Learn More
GSDMB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12885-1-AP-150UL
This item is not returnable.
View return policy
Description
The GSDMB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GSDMB. This antibody recognizes human antigen. The GSDMB antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| GSDMB | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 55876 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |