missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17032-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the guanine nucleotide-binding protein-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.Specifications
| GRK5 | |
| Polyclonal | |
| Unconjugated | |
| GRK5 | |
| GPRK5, GRK5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14773, 2869, 59075 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P34947, Q62833, Q8VEB1 | |
| GRK5 | |
| GRK5 Fusion Protein Ag10511 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction