missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIPAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15806-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 97 kDa, and 15806-1-AP specifically recognizes the 97 kDa protein.
GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain.Specifications
| GRIPAP1 | |
| Polyclonal | |
| Unconjugated | |
| GRIPAP1 | |
| GRASP 1, GRIP1 associated protein 1, GRIPAP1, KIAA1167 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116493, 54645, 56850 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4V328, Q8VD04, Q9JHZ4 | |
| Gripap1 | |
| GRIPAP1 Fusion Protein Ag8540 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction