Learn More
GRIPAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15806-1-AP-150UL
 View more versions of this product
Product Code. 16865154
  • 4836.10 NOK / 150µL
Description
The MW of this protein is 97 kDa, and 15806-1-AP specifically recognizes the 97 kDa protein.
GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain.
Specifications
Specifications
GRIPAP1
Polyclonal
Unconjugated
GRIPAP1
GRASP 1, GRIP1 associated protein 1, GRIPAP1, KIAA1167
Rabbit
Antigen Affinity Chromatography
RUO
116493, 54645, 56850
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4V328, Q8VD04, Q9JHZ4
Gripap1
GRIPAP1 Fusion Protein Ag8540
150 μL
Primary
Human, Mouse, Rat