Learn More
Checkers Grip Guard™ 5 Channel Lightweight Cable Protector
Product Code. 17782418
Change view
Click to view available options
Color:
Yellow, Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17782418 Yellow, Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17782418 Supplier Checkers Supplier No. PGGS0002YG
  • ONLINE EXCLUSIVE PRICE
    2678.21 NOK
    2175.67 NOK / Pack of 1
    Save 502.54 NOK
    18% Off
Estimated Shipment: 21-12-2026
Lightweight, durable 5-channel cable protector.
  • The Grip Guard™ 5-Channel Cable Protector includes many patent-pending features, including our 'Gripper Connectors' and new lightweight design
  • At a weight of only 58 kg, Grip Guard is up to 60% lighter than other leading polyurethane 5-channel cable protectors, and up to 75% lighter than other rubber 5-channel cable protectors on the market
  • The Gripper Connector is ergonomically designed to fit your hand, which allows for easy handling and transport
  • This connector also locks multiple cable protectors tightly together to provide a safe crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines
  • This modular interlocking design is ideal for use in public entertainment venues, light commercial, and utility applications
  • Grip Guard™ Cable Protectors are made from maintenance free polyurethane
  • When compared to other rubber manufactured cable protectors, the Grip Guard™ 5-Channel is up to 75% lighter without compromising the required strength and durability
Specifications
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 42 cm
Color Yellow, Gray
Material Polyurethane
Type Cable Protector