Learn More
SPEX Certiprep™ Stainless Steel Grinding Vial Set
Product Code. 12584316 Shop All SPEX Certiprep Products
Change view
Click to view available options
Volume (Metric):
0.1 to 0.5 g
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12584316 0.1 to 0.5 g 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12584316 Supplier SPEX Certiprep™ Supplier No. 6781S
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Used to grind hard samples like bone and circuit boards
  • For use when contact with plastic is not recommended.
  • Set includes stainless steel impactor, stainless steel center cylinder, two end plugs, and a pack of 10 O-rings.
  • Sample grinding capacity of 0.1 gram - 5.0 gram.
Specifications
For Use With (Equipment) Freezer Mills
Material Stainless Steel
Volume (Metric) 0.1 to 0.5 g