Learn More
Retsch™ Grinding Jar
Shop All Retsch Products
Click to view available options
Material:
Hardened Steel
Stainless Steel
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Description
- Screw-top
- For convenient leak-proof operation
- Constructed of stainless steel or hardened steel
- Performs radial oscillations in a horizontal position
Specifications
Specifications
| Product Type | Grinding Jar |
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | CryoMill |
| Material | Stainless Steel |