Learn More
IKA™ Grinding Chambers
Product Code. 10163331 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
80 mL
250 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10163331 80 mL Each
10021081 250 mL Each
2 options
Product Code. 10163331
Brand: IKA™ 0002983100
Only null left
Specifications
Product Type Grinding Chamber
Capacity (Metric) 80 mL
For Use With (Equipment) Compatible with A11 basic analytical mill
Material Fiberglass-reinforced ETFE
Material (Secondary) Stainless Steel Inlet
Model A11.5