Learn More
Abnova™ GRIK2 (Human) Recombinant Protein
Product Code. 16125761
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16125761 25 μg 25µg
16115761 10 μg 10µg
2 options
Product Code. 16125761
Brand: Abnova™ H00002898P01.25ug
Human GRIK2 full-length ORF ( AAH37954, 1 a.a. - 353 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MKIIFPILSNPVFRRTVKLLLCLLWIGYSQGTTHVLRFGGIFEYVESGPMGAEELAFRFAVNTINRNRTLLPNTTLTYDTQKINLYDSFEASKKACDQLSLGVAAIFGPSHSSSANAVQSICNALGVPHIQTRWKHQVSDNKDSFYVSLYPDFSSLSRAILDLVQFFKWKTVTVVYDDSTGLIRLQELIKAPSRYNLRLKIRQLPADTKDAKPLLKEMKRGKEFHVIFDCSHEMAAGILKQALAMGMMTEYYHYIFTTLDLFALDVEPYRYSGVNMTGFRILNTENTQVSSIIEKWSMERLQAPPKPDSGLLDGFMTMEFHSCCPGWSAMARSHLTAASASWVQAILLPQPPE
Specifications
Accession Number AAH37954
Gene ID (Entrez) 2898
Name glutamate receptor, ionotropic, kainate 2
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias EAA4, GLR6, GLUK6, GLUR6, MGC74427, MRT6
Gene Symbol GRIK2
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer