Learn More
Vitlab™ Griffin Beaker
Product Code. 10270981
Click to view available options
Capacity (Metric):
50 mL
150 mL
250 mL
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 10270981
Brand: Vitlab™ 61003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make your job easier and safer while handling a wide range of liquids with these beakers. VITLAB™ Griffin Beakers feature an easy to read, embossed scale.
  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C
Specifications
Color Crystal Clear
Diameter (Metric) Outer 77 mm
Capacity (Metric) 250 mL
Material PMP
Height (Metric) 95 mm
Quantity 12 Pk.
Type Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Red Printed Scale
Style Griffin