Learn More
Gilac™ Grid
Product Code. p-200009557
Click to view available options
For Use With (Equipment):
10 L Flat Tray
2 L Flat Tray
3 L Flat Tray
5 L Flat Tray
600 x 400 Container
8 L Flat Tray
Unit Size:
Each
Product Code. 17958781
Brand: Gilac™ G102621
Only null left
Add to basket
Add to basket
Grid is highly durable and shock resistant; it avoids any contact between food and exudates.
  • Grid for flat trays.
  • Highly durable and shock resistant.
  • Enables the use of flat dishes as defrosting trays by separating the ice water from the product being defrosted.
Specifications
Product Type Container Grid
Material Polyethylene
For Use With (Equipment) 2 L Flat Tray