Learn More
Gilac™ Grid
Product Code. 17988701
Change view
Click to view available options
For Use With (Equipment):
10 L Flat Tray
2 L Flat Tray
3 L Flat Tray
5 L Flat Tray
600 x 400 Container
8 L Flat Tray
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17988701 5 L Flat Tray Each
17958781 2 L Flat Tray Each
17958421 3 L Flat Tray Each
17978701 8 L Flat Tray Each
17948561 10 L Flat Tray Each
17928711 600 x 400 Container Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17988701 Supplier Gilac™ Supplier No. G100121
  • ONLINE EXCLUSIVE PRICE
    41.58 NOK
    29.36 NOK / Pack of 1
    Save 12.22 NOK
    29% Off
Estimated Shipment: 04-12-2026
Grid is highly durable and shock resistant; it avoids any contact between food and exudates.
  • Grid for flat trays.
  • Highly durable and shock resistant.
  • Enables the use of flat dishes as defrosting trays by separating the ice water from the product being defrosted.
Specifications
Product Type Container Grid
Material Polyethylene
For Use With (Equipment) 5 L Flat Tray