Description
- Constructed of fibreboard/cardboard
- Water-repellent for longer durability
- Can be used with a large range of grid dividers
Specifications
Specifications
| Color | Green |
| Material | Cardboard |
| Shape | Square |
| Type | Cryo Box |
| Dimensions (L x W x H) | 136 x 136 x 100 mm |