Learn More
Socorex™ Green Capillary Tube
Specifications
Specifications
| Volume (Metric) | 10 to 50 μL |
| Color | Green |
| Quantity | 200/Pk. |
| Product Type | Capillary Tube |
| For Use With (Equipment) | Acura 846 Positive Displacement Pipette |
| Material | Glass |