missing translation for 'onlineSavingsMsg'
Learn More

GRB2 Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 1C5D11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL594-66880-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17701439

  • 3499.43 NOK / 100µL
Estimated Shipment: 28-09-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GRB2 Monoclonal antibody specifically detects GRB2 in Human, Mouse, Rat samples. It is validated for IF
Specifications

Specifications

GRB2
Monoclonal
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Mouse
100 μL
2885
Store at -20°C
Liquid
Immunofluorescence
1C5D11
CoraLite 594
BC010915
Protein G purification
Primary
Human, Mouse, Rat
Antibody
IgG1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers