Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
Description
- Acc. to DIN 12 871
- Durometer 40 Shore A
- Available in a wide range of sizes
Specifications
Specifications
| Height (Metric) | 20mm |
| Quantity | 1/Ea. |
| Color | Gray |
| Diameter (Metric) Bottom | 14 mm |
| Diameter (Metric) Top | 18 mm |
| Material | Natural Rubber |
| Product Type | Tapered Stopper |