Learn More
Trajan Scientific and Medical™ Graphite/Vespel Ferrule
Click to view available options
Diameter (Metric) Inside:
0.3 mm
0.4 mm
Unit Size:
Pack of 10
Description
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms a strong grip with the capillary column
Specifications
Specifications
| Type | 2-hole Ferrule |
| Length (Metric) | 3.7 mm |
| Width (Metric) | 3.2 mm |
| For Use With (Equipment) | 1/16 in. Fittings |
| Material | Graphite, Vespel |
| Quantity | 10 Pack |
| Diameter (Metric) Inside | 0.4 mm |